Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C32615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32615G 5 g
C32611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32615G Supplier TCI America Supplier No. C32615G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3260-93-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00179306
InChI Key NKVUYEGKHRDEBB-UHFFFAOYSA-N
Synonym 3-Chloro-o-anisic Acid
PubChem CID 6423044
IUPAC Name 3-chloro-2-methoxybenzoic acid
SMILES COC1=C(C=CC=C1Cl)C(O)=O

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-methoxybenzoic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.