Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Chloro-2-methyl-1-propene

Catalog No. 501859586
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-9586 1 L
50-185-9585 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-9586 Supplier Sigma Aldrich Supplier No. 1080301L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Chloro-2-methyl-1-propene
TRUSTED_SUSTAINABILITY

Specifications

CAS 563-47-3
Melting Point -80°C (lit.)
Density 0.925 g/mL (at 20°C (literature))
Boiling Point 71°C to 72°C (lit.)
Molecular Formula C4H7Cl
Refractive Index n20/D 1.427 (literature)
Linear Formula CH2=C(CH3)CH2Cl
MDL Number MFCD00000953
Quantity 1 L
Synonym beta-Methallyl chloride
Molecular Weight (g/mol) 90.55
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.