Alfa Aesar™ 3-Chloro-2-methylanisole, 97%

Manufacturer: Alfa Aesar™ B2220322

Quantity 100g

 View more versions of this product

Catalog No. AAB2220322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3260-88-6
    Molecular Formula C8H9ClO
    Formula Weight 156.61
    MDL Number MFCD00070772
    UN Number UN1993
    Beilstein 1862220
    Synonym 2-Chloro-6-methoxytoluene; 2-Methoxy-6-chlorotoluene
    Chemical Name or Material 3-Chloro-2-methylanisole

    Specifications

    Formula Weight 156.61
    TSCA TSCA
    Flash Point 92°C (197°F)
    Melting Point −3°C
    Quantity 100g
    Molecular Formula C8H9ClO
    MDL Number MFCD00070772
    CAS 3260-88-6
    UN Number UN1993
    Beilstein 1862220
    Synonym 2-Chloro-6-methoxytoluene; 2-Methoxy-6-chlorotoluene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Boiling Point 212° to 215°C
    Chemical Name or Material 3-Chloro-2-methylanisole
    Assay 97%

    Description

    Applications
    Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit