Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-methylbenzonitrile, 97%

Catalog No. AAL1104806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1104806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1104806 Supplier Thermo Scientific Chemicals Supplier No. L1104806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 54454-12-5 | C8H6ClN | 151.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54454-12-5
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00045598
InChI Key FKFZTNLSUJCIMG-UHFFFAOYSA-N
Synonym 2-methyl-3-chlorobenzonitrile, pubchem23847, 3-chloro-o-tolunitrile, acmc-1aqtx, 6-chloro-2-cyanotoluene, 2-chloro-6-cyanotoluene, 2-methyl-3-chloro-benzonitrile, 3-chloro-2-methyl-benzonitrile, benzonitrile,3-chloro-2-methyl, benzonitrile, 3-chloro-2-methyl
PubChem CID 521508
IUPAC Name 3-chloro-2-methylbenzonitrile
SMILES CC1=C(C=CC=C1Cl)C#N

Specifications

Melting Point 20°C to 23°C
Quantity 5 g
Refractive Index 1.553
UN Number UN3439
Beilstein 2613370
Boiling Point 120°C (22 mmHg)
Formula Weight 151.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-methylbenzonitrile
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.