Learn More
Medchemexpress LLC 3-Chloro-2-pyrazinamine | 6863-73-6 | 129.55 | 5 G
SDP
Catalog No. 5000446411
Encompass_Preferred
3-Chloro-2-pyrazinamine is an off-white to light yellow solid biochemical reagent. It is suitable for use as a biological material or organic compound in various life science-related research applications. This product is intended for research use only.
  • Suitable for life science research
  • Off-white to light yellow solid
  • Store at 4°C, protected from light (solid)
  • Store at -80°C for 6 months or -20°C for 1 month in solvent, protected from light
Catalog No. 50-004-46411 Supplier Medchemexpress LLC Supplier No. HYW0074995G
3-Chloro-2-pyrazinamine is an off-white to light yellow solid biochemical reagent. It is suitable for use as a biological material or organic compound in various life science-related research applications. This product is intended for research use only.
  • Suitable for life science research
  • Off-white to light yellow solid
  • Store at 4°C, protected from light (solid)
  • Store at -80°C for 6 months or -20°C for 1 month in solvent, protected from light