Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-2-pyridinecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C23421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23421G 1 g
C23425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23421G Supplier TCI America Supplier No. C23421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57266-69-0
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD01646325
InChI Key XTMUXJBJCMRWPG-UHFFFAOYSA-N
Synonym 3-chloropicolinic acid, 3-chloro-2-pyridinecarboxylic acid, 3-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 3-chloro, 3-chloropyridine-2-carboxylicacid, 3-chloro-2-picolinicacid, 3-chloro-pyridine-2-carboxylic acid, chloropicolinic acid, pubchem12918, 3-chloro picolinic acid
PubChem CID 824632
IUPAC Name 3-chloropyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)Cl

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 1 g
Formula Weight 157.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-pyridinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.