Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-3-methylhexane, 97%, Thermo Scientific™

Catalog No. AAB2027714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2027714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2027714 Supplier Thermo Scientific Chemicals Supplier No. B2027714

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43197-78-0
Molecular Formula C7H15Cl
Molecular Weight (g/mol) 134.65
MDL Number MFCD00060778
InChI Key UTKDCNDVTOWUHW-UHFFFAOYNA-N
Synonym hexane, 3-chloro-3-methyl, acmc-20an6z, 3-chlor-3-methyl-hexan, hexane,3-chloro-3-methyl
PubChem CID 142659
IUPAC Name 3-chloro-3-methylhexane
SMILES CCCC(C)(Cl)CC

Specifications

Density 0.879
Boiling Point 135°C
Flash Point 35°C (95°F)
Refractive Index 1.4265
Linear Formula CH3CH2CH2CCl(CH3)CH2CH3
Quantity 25 g
UN Number UN1993
Formula Weight 134.65
Percent Purity 97%
Chemical Name or Material 3-Chloro-3-methylhexane
Hazard Category H226
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.