Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 3-Chloro-4-fluorobenzaldehyde, 34328-61-5, MFCD00011735, 25g

Catalog No. 501473332
Encompass_Preferred

Molecular Formula C7H4ClFO, Purity 97%, Molecular Weight 158.56, Melting Point 28-30°, Boiling Point 66-72/2mm

Catalog No. 50-147-3332 Supplier Matrix Scientific Supplier No. 00250925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C7H4ClFO, Purity 97%, Molecular Weight 158.56, Melting Point 28-30°, Boiling Point 66-72/2mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.