Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 3-Chloro-4-Fluorophenylmagnesium Bromide Solution
missing translation for 'brandsLinkText'
Description
Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.
Spécifications
Spécifications
| CAS | 82297-90-3 |
| Density | 0.972 g/mL at 25°C |
| Boiling Point | 65°C |
| Molecular Formula | C6H3BrCl2Mg |
| Linear Formula | Cl2C6H3MgBr |
| Quantity | 50 mL |
| MDL Number | MFCD01319892 |
| Molecular Weight (g/mol) | 250.2 g/mol |
| Biological Activity | Respiratory System |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'