Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-Chloro-4-hydroxybenzaldehyde, 98%, [2420-16-8], Y15231 156.566 C7H5ClO2

Catalog No. 501456401
Encompass_Preferred

Compound: 3-Chloro-4-hydroxybenzaldehyde

CAS#: 2420-16-8

MDL#: not listed

Background: A chlorinated hydroxybenzaldehyde used as a common building block in pharmaceutical research. The chlorine substituent offers a reactive site for cross-coupling, the phenolic hydroxyl allows for etherification, and the aldehyde permits further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-145-6401 Supplier Frontier Specialty Chemicals Supplier No. Y152315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Chloro-4-hydroxybenzaldehyde

CAS#: 2420-16-8

MDL#: not listed

Background: A chlorinated hydroxybenzaldehyde used as a common building block in pharmaceutical research. The chlorine substituent offers a reactive site for cross-coupling, the phenolic hydroxyl allows for etherification, and the aldehyde permits further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.