Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-hydroxybenzaldehyde, 98%

Catalog No. AAA1751514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1751514 25 g
AAA1751506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1751514 Supplier Thermo Scientific Chemicals Supplier No. A1751514
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2420-16-8 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2420-16-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00016981
InChI Key VGSOCYWCRMXQAB-UHFFFAOYSA-N
Synonym benzaldehyde, 3-chloro-4-hydroxy, 4-hydroxy-3-chlorobenzaldehyde, 3-chloro-4-hydroxy-benzaldehyde, benzaldehyde, 3-chloro-4-hydroxy-8ci 9ci, pubchem7764, acmc-1crqk, 3-chlor-4-hydroxybenzaldehyde, 5-chloro-4-hyroxybenzaldehyde, 5-chloro-4-hydroxybenzaldehyde, 3-chloro-4-hydroxy benzaldehyde
PubChem CID 17022
IUPAC Name 3-chloro-4-hydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C=C1Cl

Specifications

Melting Point 128°C to 132°C
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.