Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-4-methoxyphenylboronic acid, 95%, Thermo Scientific™

Catalog No. AC427920050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427920050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427920050 Supplier Thermo Scientific Chemicals Supplier No. 427920050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175883-60-0
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD04039888
InChI Key VUTJHOWRPUPHIG-UHFFFAOYSA-N
Synonym 3-chloro-4-methoxyphenyl boronic acid, 3-chloro-4-methoxybenzeneboronic acid, 4-borono-2-chloroanisole, 3-chloro-4-methoxyphenyl boranediol, boronic acid, 3-chloro-4-methoxyphenyl, akos brn-0653, 3-chloro-4, acmc-1c1io, ksc489k9h, 3-chloro-4-methoxyphenyl boronicacid
PubChem CID 3247522
IUPAC Name (3-chloro-4-methoxyphenyl)boronic acid
SMILES COC1=C(Cl)C=C(C=C1)B(O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 240°C to 250°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(Cl)B(OH)2
Quantity 5 g
Formula Weight 186.4
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-methoxyphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.