Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-4-methylbenzaldehyde, 97%

Catalog No. AC436690050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC436690050 5 g
AC436690010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436690050 Supplier Thermo Scientific Chemicals Supplier No. 436690050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3411-03-8 | C8H7ClO | 154.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3411-03-8
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
InChI Key OJFQNFVCICBYQC-UHFFFAOYSA-N
Synonym 3-chloro-4-methylbenzaldehyde, isobornylcyclohexanol, pubchem4183, 2-chloro-4-formyltoluene, ksc497m3l, ojfqnfvcicbyqc-uhfffaoysa-n, benzaldehyde, 3-chloro-4-methyl, 3-5,6,6-trimethyl-2-norbornyl-cyclohexanol
PubChem CID 12423085

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C to 31°C
Color White or Colorless
Boiling Point 232°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 154.6
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-chloro-4-methylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.