Learn More
Medchemexpress LLC (3-chloro-4-methylphenyl)-[4-(4-nitrophenyl)piperazin-1-yl]methanone | 431996-53-1 | MFCD03058358 | 99.3% | 359.81 g·mol⁻1 | C18H18ClN3O3 | 10MG
SDP
Catalog No. 5000239198
Encompass_Preferred
Filastatin is a research-grade small molecule that inhibits Candida albicans filamentation and reduces fungal adhesion. It blocks the yeast-to-hyphal morphological transition and has been shown to inhibit adhesion to polystyrene and human cells, with an IC50 of approximately 3 μM in a GFP-based adhesion assay.
  • Potent inhibitor of Candida albicans filamentation (IC50 ≈ 3 μM).
  • Reduces fungal adhesion to polystyrene and human cells.
  • Blocks yeast-to-hyphal morphological transition.
  • High chemical purity suitable for research applications.
  • Supplied as a solid for easy handling and storage.
Catalog No. 50-002-39198 Supplier Medchemexpress LLC Supplier No. HY12470110MG
Filastatin is a research-grade small molecule that inhibits Candida albicans filamentation and reduces fungal adhesion. It blocks the yeast-to-hyphal morphological transition and has been shown to inhibit adhesion to polystyrene and human cells, with an IC50 of approximately 3 μM in a GFP-based adhesion assay.
  • Potent inhibitor of Candida albicans filamentation (IC50 ≈ 3 μM).
  • Reduces fungal adhesion to polystyrene and human cells.
  • Blocks yeast-to-hyphal morphological transition.
  • High chemical purity suitable for research applications.
  • Supplied as a solid for easy handling and storage.