Learn More
Medchemexpress LLC (3-chloro-4-methylphenyl)-[4-(4-nitrophenyl)piperazin-1-yl]methanone | 431996-53-1 | MFCD03058358 | 99.3% | 359.81 g/mol | C18H18ClN3O3 | 100MG
SDP
Catalog No. 5000237356
Encompass_Preferred
Filastatin is a small-molecule antifungal research reagent that inhibits Candida albicans filamentation and adhesion. It blocks the yeast-to-hyphal morphological transition and reduces fungal attachment to abiotic and human surfaces, showing activity at low micromolar concentrations.
  • Inhibits Candida albicans filamentation.
  • Reduces fungal adhesion to polystyrene and human cells.
  • Active at low micromolar concentrations (IC50 ~3 μM).
  • High reported purity suitable for research applications (≈99.3%).
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-37356 Supplier Medchemexpress LLC Supplier No. HY124701100MG
Filastatin is a small-molecule antifungal research reagent that inhibits Candida albicans filamentation and adhesion. It blocks the yeast-to-hyphal morphological transition and reduces fungal attachment to abiotic and human surfaces, showing activity at low micromolar concentrations.
  • Inhibits Candida albicans filamentation.
  • Reduces fungal adhesion to polystyrene and human cells.
  • Active at low micromolar concentrations (IC50 ~3 μM).
  • High reported purity suitable for research applications (≈99.3%).
  • Available in multiple pack sizes for experimental flexibility.