Learn More
Medchemexpress LLC (3-chloro-4-methylphenyl)-[4-(4-nitrophenyl)piperazin-1-yl]methanone | 431996-53-1 | MFCD03058358 | 99.3% | 359.81 g/mol | C18H18ClN3O3 | 5MG
SDP
Catalog No. 5000239225
Encompass_Preferred
Filastatin is a small-molecule antifungal research compound (CAS 431996-53-1) that inhibits Candida albicans filamentation and adhesion. It suppresses the yeast-to-hyphal morphological transition, reduces biofilm formation, and blocks adhesion of multiple pathogenic Candida species (reported IC50 ≈ 3 μM in a GFP-based adhesion assay).
  • Inhibits Candida albicans filamentation and adhesion.
  • Reported IC50 ≈ 3 μM in a GFP-based adhesion assay.
  • Reduces biofilm formation and hyphal morphogenesis.
  • Does not affect A549 cell viability at high concentrations in vitro.
  • Supplied as a solid, light yellow to yellow powder, 5 mg pack, purity 99.27%.
Catalog No. 50-002-39225 Supplier Medchemexpress LLC Supplier No. HY1247015MG
Filastatin is a small-molecule antifungal research compound (CAS 431996-53-1) that inhibits Candida albicans filamentation and adhesion. It suppresses the yeast-to-hyphal morphological transition, reduces biofilm formation, and blocks adhesion of multiple pathogenic Candida species (reported IC50 ≈ 3 μM in a GFP-based adhesion assay).
  • Inhibits Candida albicans filamentation and adhesion.
  • Reported IC50 ≈ 3 μM in a GFP-based adhesion assay.
  • Reduces biofilm formation and hyphal morphogenesis.
  • Does not affect A549 cell viability at high concentrations in vitro.
  • Supplied as a solid, light yellow to yellow powder, 5 mg pack, purity 99.27%.