Learn More
Medchemexpress LLC 3-chloro-4-methylphenylboronic acid | 175883-63-3 | MFCD04039010 | 99.9% | 170.40 g/mol | C7H8BClO2 | 5 G
SDP
Catalog No. 5000299989
Encompass_Preferred
(3-Chloro-4-methylphenyl)boronic acid is an aromatic boronic acid used as a building block in organic synthesis and life-science research. It is commonly employed in Suzuki-Miyaura cross-coupling reactions to install aryl groups. The material is supplied as a white to off-white solid with high purity for laboratory use.
  • Aromatic boronic acid suitable for Suzuki-Miyaura cross-coupling.
  • High purity for consistent reaction performance.
  • Available in small gram-scale packages for laboratory applications.
  • White to off-white solid for straightforward handling and storage.
  • Molecular formula C7H8BClO2 and molecular weight 170.40 g/mol.
Catalog No. 50-002-99989 Supplier Medchemexpress LLC Supplier No. HYW0169635G
Add to Cart
Add to Cart
(3-Chloro-4-methylphenyl)boronic acid is an aromatic boronic acid used as a building block in organic synthesis and life-science research. It is commonly employed in Suzuki-Miyaura cross-coupling reactions to install aryl groups. The material is supplied as a white to off-white solid with high purity for laboratory use.
  • Aromatic boronic acid suitable for Suzuki-Miyaura cross-coupling.
  • High purity for consistent reaction performance.
  • Available in small gram-scale packages for laboratory applications.
  • White to off-white solid for straightforward handling and storage.
  • Molecular formula C7H8BClO2 and molecular weight 170.40 g/mol.