Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-methylpyridine, 95%, Thermo Scientific™

Catalog No. AAH6162606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6162606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6162606 Supplier Thermo Scientific Chemicals Supplier No. H6162606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72093-04-0
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD04114245
InChI Key VLNXNJMHLZFECV-UHFFFAOYSA-N
Synonym 3-chloro-4-picoline, pyridine, 3-chloro-4-methyl, 3-chloro-4-methyl-pyridine, pubchem14323, acmc-1bl3c, 3-chloro-4-methylpyridine?, ksc494c8t, 3-chloro-4-methylpyridine
PubChem CID 2762925
IUPAC Name 3-chloro-4-methylpyridine
SMILES CC1=C(Cl)C=NC=C1

Specifications

Color Colorless to Brown
Quantity 5 g
Formula Weight 127.57
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-methylpyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.