Learn More
Sigma Aldrich 3-Chloro-4-(trifluoromethyl)pyridine-2-carboxylic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Precursor involved in the synthesis of pharmacologically active molecules including:
- Selective orally active s1p1 agonists
- Water-soluble prodrugs of triazole cs-758 with antifungal activity
- Fostriecin analogs as antitumor agentsreactant involved in:
- Phosphitylation of alcohols
- Deallylation for the synthesis of nucleoside phosphoramidite
- Posphitylation and stereoselective pudovik rearrangement
Specifications
Specifications
| Molecular Formula | C7H3ClF3NO2 |
| Linear Formula | C7H3ClF3NO2 |
| MDL Number | MFCD14581664 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 225.55 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.