Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-fluorobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. C33305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33305G 5 g
C33301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33305G Supplier TCI America Supplier No. C33305G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 493038-93-0
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD04038259
InChI Key GWPLYSLPTCBNDL-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorophenylacetonitrile, 2-3-chloro-5-fluorophenyl acetonitrile, 3-chloro-5-fluorobenzyl cyanide, 3-chloro-5-fluorophenyl acetonitrile, 2-3-chloro-5-fluoro-phenyl acetonitrile, pubchem4280, ksc494a3d, 3-chloro-5-fluorobenzylcyanide, 2-5-chloro-3-fluorophenyl ethanenitrile
PubChem CID 2734836
IUPAC Name 2-(3-chloro-5-fluorophenyl)acetonitrile
SMILES C1=C(C=C(C=C1F)Cl)CC#N

Specifications

Color Yellow
Quantity 5 g
UN Number 3276
Formula Weight 169.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-5-fluorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.