Learn More
Selleck Chemical LLC 3-chloro-5-hydroxybenzoic Acid S5400-1g
SDP
Catalog No. 503110262
Encompass_Preferred
3-chloro-5-hydroxybenzoic Acid is a selective agonist of the lactate receptor GPR81
Catalog No. 50-311-0262 Supplier Selleck Chemical LLC Supplier No. S54001G
3-chloro-5-hydroxybenzoic Acid is a selective agonist of the lactate receptor GPR81