Learn More
Medchemexpress LLC 3-chloro-5-hydroxybenzoic acid | 53984-36-4 | MFCD04114327 | 99.5% | 172.57 g·mol⁻¹ | C7H5ClO3 | 100 MG
SDP
Catalog No. 5000300909
Encompass_Preferred
3-Chloro-5-hydroxybenzoic acid (CAS 53984-36-4) is a small-molecule agonist of the lactate receptor GPR81 (HCAR1) used in research to probe receptor signaling and metabolic effects. It is supplied with high reported purity and in formats suitable for biochemical screening and in vivo testing.
  • Potent GPR81 agonist, human EC50 = 16 μM.
  • Reported purity 99.54%.
  • Molecular formula C7H5ClO3, molecular weight 172.57 g·mol⁻¹.
  • Available as solid or DMSO solution for screening.
  • Suitable for in vitro and in vivo metabolic and lipolysis studies.
Catalog No. 50-003-00909 Supplier Medchemexpress LLC Supplier No. HYW016868100MG
3-Chloro-5-hydroxybenzoic acid (CAS 53984-36-4) is a small-molecule agonist of the lactate receptor GPR81 (HCAR1) used in research to probe receptor signaling and metabolic effects. It is supplied with high reported purity and in formats suitable for biochemical screening and in vivo testing.
  • Potent GPR81 agonist, human EC50 = 16 μM.
  • Reported purity 99.54%.
  • Molecular formula C7H5ClO3, molecular weight 172.57 g·mol⁻¹.
  • Available as solid or DMSO solution for screening.
  • Suitable for in vitro and in vivo metabolic and lipolysis studies.