Learn More
Medchemexpress LLC 3-chloro-5-hydroxybenzoic acid | 53984-36-4 | MFCD04114327 | 99.5% | 172.57 g/mol | C7H5ClO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01686850MG
Encompass_Preferred
3-chloro-5-hydroxybenzoic acid is a benzoic acid derivative supplied as a research-grade reagent. It is reported as a selective, orally active GPR81 (HCAR1) agonist used in studies of lipolysis and metabolic disease. Manufacturer documentation includes a certificate of analysis and a safety data sheet for quality and safety reference.
  • High purity (listed at 99.54%) for biochemical assays.
  • Molecular formula C7H5ClO3 and molecular weight 172.57 g/mol.
  • Reported GPR81 (lactate receptor) agonist activity (EC50 ≈16 μM).
  • Provided with COA and SDS for traceability and safety compliance.
  • Off-white solid form suitable for storage and handling in research labs.
Catalog No. 50-003-01936