Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. C12575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12575G Supplier TCI America Supplier No. C12575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic reagent for the preparation of polycyclic aromatic heterocycles.

Chemical Identifiers

CAS 74115-12-1
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006379
InChI Key TUIDQYRZDZRHPQ-UHFFFAOYSA-N
Synonym 5-chloro-3-hydroxypyridine, 5-chloro-3-pyridinol, 3-chloro-5-hydroxypyridine, 3-pyridinol, 5-chloro, 5chloro-3-pyridinol, pubchem6628, 3-chloro-5-pyridinol, acmc-209otf, 5-chloranylpyridin-3-ol, 3chloro-5-hydroxypyridine
PubChem CID 582305
IUPAC Name 5-chloropyridin-3-ol
SMILES C1=C(C=NC=C1Cl)O

Specifications

Melting Point 161°C
Quantity 5 g
Formula Weight 129.54
Percent Purity ≥98.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-5-hydroxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.