Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-6-hydrazinopyridazine, 98%

Catalog No. AAL0612106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0612106 5 g
AAL0612103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0612106 Supplier Thermo Scientific Chemicals Supplier No. L0612106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17284-97-8 | C4H5ClN4 | 144.562 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17284-97-8
Molecular Formula C4H5ClN4
Molecular Weight (g/mol) 144.562
MDL Number MFCD00051740
InChI Key FXYQRYGWWZKUFV-UHFFFAOYSA-N
Synonym 3-chloro-6-hydrazinopyridazine, 3-chloro-6-hydrazinylpyridazine, 3-chloropyridazin-6-yl hydrazine, 1-6-chloropyridazin-3-yl hydrazine, 6-chloro-pyridazin-3-yl-hydrazine, 3-chloro-6-hydrazino-pyridazine, 6-chloropyridazin-3-yl hydrazine, 3 2h-pyridazinone, 6-chloro-, hydrazone, 6-chloropyridazine-3-ylhydrazine, 6-chloro-3-pyridazinyl hydrazine
PubChem CID 100787
IUPAC Name (6-chloropyridazin-3-yl)hydrazine
SMILES C1=CC(=NN=C1NN)Cl

Specifications

Melting Point 134°C to 140°C
Quantity 5 g
Beilstein 125591
Formula Weight 144.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-6-hydrazinopyridazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.