Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-6-methylpyridazine, 97%, Thermo Scientific™

Catalog No. AAA1348006
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348006 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1348006 Supplier Thermo Scientific Chemicals Supplier No. A1348006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1121-79-5
Molecular Formula C5H5ClN2
Quantity 5g
MDL Number MFCD00052905
Beilstein 108655
Synonym 3-chloro-6-methyl-pyridazine, 3-chloro-6-methyl pyridazine, pyridazine, 3-chloro-6-methyl, 6-chloro-3-methylpyridazine, pubchem10467, 3-methyl-6-chloropyridazine, 6-methyl-3-chloropyridazine, 3-chloro-6-methyl-1,2-diazine, 3-chloro-6-methylpyridazine
InChI Key PRORLQAJNJMGAR-UHFFFAOYSA-N
SMILES CC1=NN=C(C=C1)Cl
IUPAC Name 3-chloro-6-methylpyridazine
Molecular Weight (g/mol) 128.559
PubChem CID 227254
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-6-methylpyridazine
Show More Show Less
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.