Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chloro-6-methylpyridazine, 97%, Thermo Scientific™

Catalog No. BTB06842EA
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BTB06842EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB06842EA Supplier Thermo Scientific Chemicals Supplier No. BTB06842EA

missing translation for 'validMainframeMsg'

Specifications

CAS 1121-79-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H5ClN2
Quantity 10g
MDL Number MFCD00052905
Synonym 3-chloro-6-methyl-pyridazine, 3-chloro-6-methyl pyridazine, pyridazine, 3-chloro-6-methyl, 6-chloro-3-methylpyridazine, pubchem10467, 3-methyl-6-chloropyridazine, 6-methyl-3-chloropyridazine, 3-chloro-6-methyl-1,2-diazine, 3-chloro-6-methylpyridazine
InChI Key PRORLQAJNJMGAR-UHFFFAOYSA-N
SMILES CC1=NN=C(C=C1)Cl
IUPAC Name 3-chloro-6-methylpyridazine
Molecular Weight (g/mol) 128.559
PubChem CID 227254
Formula Weight 128.56
Percent Purity 97%
Chemical Name or Material 3-chloro-6-methylpyridazine
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.