Learn More
Medchemexpress LLC HY-W041171 250mg Medchemexpress, 3-Chloro-L-tyrosine CAS:7423-93-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW041171250MG
Encompass_Preferred
Medchemexpress, HY-W041171 250mg 3-Chloro-L-tyrosine CAS:7423-93-0 3-Chloro-L-tyrosine is a specific marker of myeloperoxidase-catalyzed oxidation, and is markedly elevated in low density lipoprotein isolated from human atherosclerotic intima. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4659