Learn More
Medchemexpress LLC 3-Chloro-N-(4-hydroxy-2-oxo-2H-chromen-3-yl)benzamide | 94205-22-8 | C16H10ClNO4 | 100 MG
SDP
Catalog No. 5000373298
Encompass_Preferred
Anticancer agent 93 is a 4-Hydroxycoumarin derivative that demonstrates efficacy against lung cancer cells. It inhibits invasion and migration by modulating epithelial-mesenchymal transition (EMT) effectors, making it a potential candidate for cancer research.
  • Inhibits A549 cell invasion by approximately 50%
  • Significantly inhibits A549 cell migration
  • Decreases lung cancer cell motility by downregulating EMT markers
  • Reduces expression of N-cadherin and E-cadherin in A549 cells
Catalog No. 50-003-73298 Supplier Medchemexpress LLC Supplier No. HY152084100MG
Anticancer agent 93 is a 4-Hydroxycoumarin derivative that demonstrates efficacy against lung cancer cells. It inhibits invasion and migration by modulating epithelial-mesenchymal transition (EMT) effectors, making it a potential candidate for cancer research.
  • Inhibits A549 cell invasion by approximately 50%
  • Significantly inhibits A549 cell migration
  • Decreases lung cancer cell motility by downregulating EMT markers
  • Reduces expression of N-cadherin and E-cadherin in A549 cells