Learn More
Medchemexpress LLC 3-Chloro-N-(4-hydroxy-2-oxo-2H-chromen-3-yl)benzamide | 94205-22-8 | C16H10ClNO4 | 25 MG
SDP
Catalog No. 5000374285
Encompass_Preferred
Anticancer agent 93 is a 4-Hydroxycoumarin derivative. It can inhibit invasion and migration of lung cancer cells by modulating the expression of epithelial-mesenchymal transition (EMT) effectors.
  • 4-Hydroxycoumarin derivative
  • Inhibits invasion and migration of lung cancer cells
  • Modulates expression of epithelial-mesenchymal transition (EMT) effectors
  • Purity of 98.01%
  • White to off-white solid appearance
  • Inhibits A549 cell invasion by approximately 50% at concentrations of 5-15 μM for 24 hours
  • Significantly inhibits A549 cell migration at 15 μM for 24-72 hours
  • Inhibits lung cancer cell motility by downregulating EMT markers at 5-15 μM for 4 hours
  • Decreases expression of N-cadherin and E-cadherin in A549 cells
Catalog No. 50-003-74285 Supplier Medchemexpress LLC Supplier No. 25MGHY152084
Anticancer agent 93 is a 4-Hydroxycoumarin derivative. It can inhibit invasion and migration of lung cancer cells by modulating the expression of epithelial-mesenchymal transition (EMT) effectors.
  • 4-Hydroxycoumarin derivative
  • Inhibits invasion and migration of lung cancer cells
  • Modulates expression of epithelial-mesenchymal transition (EMT) effectors
  • Purity of 98.01%
  • White to off-white solid appearance
  • Inhibits A549 cell invasion by approximately 50% at concentrations of 5-15 μM for 24 hours
  • Significantly inhibits A549 cell migration at 15 μM for 24-72 hours
  • Inhibits lung cancer cell motility by downregulating EMT markers at 5-15 μM for 4 hours
  • Decreases expression of N-cadherin and E-cadherin in A549 cells