Learn More
Medchemexpress LLC 3-Chloro-N-(4-hydroxy-2-oxo-2H-chromen-3-yl)benzamide | 94205-22-8 | C16H10ClNO4 | 5 MG
SDP
Catalog No. 5000372446
Encompass_Preferred
Anticancer agent 93 is a 4-Hydroxycoumarin derivative that inhibits invasion and migration of lung cancer cells by modulating the expression of epithelial-mesenchymal transition (EMT) effectors. In vitro, it significantly inhibits A549 cell invasion and migration, and lung cancer cell motility by downregulating EMT markers.
  • Acts as a 4-hydroxycoumarin derivative
  • Inhibits invasion and migration of lung cancer cells
  • Modulates the expression of epithelial-mesenchymal transition (EMT) effectors
  • Significantly inhibits A549 cell invasion and migration in vitro
  • Downregulates EMT markers to reduce lung cancer cell motility
Catalog No. 50-003-72446 Supplier Medchemexpress LLC Supplier No. HY1520845MG
Anticancer agent 93 is a 4-Hydroxycoumarin derivative that inhibits invasion and migration of lung cancer cells by modulating the expression of epithelial-mesenchymal transition (EMT) effectors. In vitro, it significantly inhibits A549 cell invasion and migration, and lung cancer cell motility by downregulating EMT markers.
  • Acts as a 4-hydroxycoumarin derivative
  • Inhibits invasion and migration of lung cancer cells
  • Modulates the expression of epithelial-mesenchymal transition (EMT) effectors
  • Significantly inhibits A549 cell invasion and migration in vitro
  • Downregulates EMT markers to reduce lung cancer cell motility