Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Chloro-N-(4-methoxyphenyl)-4-((2-methyl-3-oxocyclopent-1-en-1-yl)amino)benzamide | 1672684-68-2 | 98.4% | 25 MG
SDP

Catalog No. 5000234175
Encompass_Preferred

MS402 is a small-molecule, BD1-selective inhibitor of BET bromodomains used in preclinical research to modulate BET-mediated transcription and Th17-driven inflammatory responses.

  • Selective BD1 bromodomain inhibition.
  • Reported Ki values for BET bromodomains, e.g., BRD4(BD1) 77 nM, BRD4(BD2) 718 nM.
  • Demonstrated activity blocking Th17 cell differentiation and ameliorating colitis in mice.
  • Purity 98.36% and molecular weight 370.83 g/mol.
  • Molecular formula C20H19ClN2O3.
  • Intended for laboratory research use only; not for human use.

Catalog No. 50-002-34175 Supplier Medchemexpress LLC Supplier No. HY12000025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MS402 is a small-molecule, BD1-selective inhibitor of BET bromodomains used in preclinical research to modulate BET-mediated transcription and Th17-driven inflammatory responses.

  • Selective BD1 bromodomain inhibition.
  • Reported Ki values for BET bromodomains, e.g., BRD4(BD1) 77 nM, BRD4(BD2) 718 nM.
  • Demonstrated activity blocking Th17 cell differentiation and ameliorating colitis in mice.
  • Purity 98.36% and molecular weight 370.83 g/mol.
  • Molecular formula C20H19ClN2O3.
  • Intended for laboratory research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.