Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-N-methylaniline 97.0+%, TCI America™
SDP

Catalog No. C18915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18915G 5 g
C189125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18915G Supplier TCI America Supplier No. C18915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7006-52-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00052014
InChI Key WFGYSQDPURFIFL-UHFFFAOYSA-N
Synonym n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
PubChem CID 138900
IUPAC Name 3-chloro-N-methylaniline
SMILES CNC1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 235°C
Quantity 5 g
Formula Weight 141.60
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-N-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.