Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chloroacetanilide, 98+%, Thermo Scientific™

Catalog No. AAA1492136
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1492136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1492136 Supplier Thermo Scientific Chemicals Supplier No. A1492136

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 588-07-8
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00016341
InChI Key MUUQHCOAOLLHIL-UHFFFAOYSA-N
Synonym 3'-chloroacetanilide, 3-chloroacetanilide, n-3-chlorophenyl acetamide, m-chloroacetanilide, acetamide, n-3-chlorophenyl, acetanilide, 3'-chloro, meta-chloroacetanilide, unii-74h09hu61j, acetic acid, amide, n 3-chlorophenyl, m-chloroacetoanilide
PubChem CID 11493
IUPAC Name N-(3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)Cl

Specifications

Melting Point 75°C to 79°C
Quantity 500 g
Beilstein 2208105
Merck Index 14,2111
Formula Weight 169.61
Percent Purity ≥98%
Chemical Name or Material 3'-Chloroacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-610-3
RTECSNumber AE1000100
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.