Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chloroacetanilide 98.0+%, TCI America™
SDP

Catalog No. C008825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C008825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C008825G Supplier TCI America Supplier No. C008825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 588-07-8
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.608
MDL Number MFCD00016341
InChI Key MUUQHCOAOLLHIL-UHFFFAOYSA-N
Synonym 3'-chloroacetanilide, 3-chloroacetanilide, n-3-chlorophenyl acetamide, m-chloroacetanilide, acetamide, n-3-chlorophenyl, acetanilide, 3'-chloro, meta-chloroacetanilide, unii-74h09hu61j, acetic acid, amide, n 3-chlorophenyl, m-chloroacetoanilide
PubChem CID 11493
IUPAC Name N-(3-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)Cl

Specifications

Melting Point 76°C
Boiling Point 172°C
Quantity 25 g
Formula Weight 169.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Chloroacetanilide
EINECSNumber (3)-0195
RTECSNumber AE1000100
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.