Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Chloroacetophenone, 98%

Catalog No. AC134840250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC134840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC134840250 Supplier Thermo Scientific Chemicals Supplier No. 134840250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 99-02-5 | C8H7ClO | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-02-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000593
InChI Key UUWJBXKHMMQDED-UHFFFAOYSA-N
Synonym 3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
PubChem CID 14933
IUPAC Name 1-(3-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC(Cl)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 227°C to 229°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5480 to 1.55
Quantity 25 g
Beilstein 07, 282
Specific Gravity 1.19
Merck Index 10, 2082
Formula Weight 154.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3'-Chloroacetophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty o
EINECSNumber 202-721-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.