Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzhydrazide, 98+%

Catalog No. AAL0107118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0107118 50 g
AAL0107109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0107118 Supplier Thermo Scientific Chemicals Supplier No. L0107118
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1673-47-8 | C7H7ClN2O | 170.596 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1673-47-8
Molecular Formula C7H7ClN2O
Molecular Weight (g/mol) 170.596
MDL Number MFCD00014757
InChI Key PHRDZSRVSVNQRN-UHFFFAOYSA-N
Synonym 3-chlorobenzhydrazide, 3-chlorobenzenecarbohydrazide, 3-chlorobenzoic hydrazide, benzoic acid, 3-chloro-, hydrazide, 3-chlorobenzoylhydrazine, 3-chlorobenzoic acid hydrazide, 3-chloro-benzoic acid hydrazide, benzoylhydrazine, m-chloro, 3-chlorobenzoic acid, hydrazide, 3-chlorobenzhyrazide
PubChem CID 74289
IUPAC Name 3-chlorobenzohydrazide
SMILES C1=CC(=CC(=C1)Cl)C(=O)NN

Specifications

Melting Point 156°C to 160°C
Quantity 50 g
Beilstein 973243
Formula Weight 170.6
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzhydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-812-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.