Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzoic acid, 97%, Thermo Scientific™

Catalog No. AAB214130I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB214130I 5000 g
AAB2141330 250 g
AAB214130B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB214130I Supplier Thermo Scientific Chemicals Supplier No. B214130I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-80-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002491
InChI Key LULAYUGMBFYYEX-UHFFFAOYSA-N
Synonym m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
PubChem CID 447
ChEBI CHEBI:49410
IUPAC Name 3-chlorobenzoic acid
SMILES C1=CC(=CC(=C1)Cl)C(=O)O

Specifications

Melting Point 155°C to 155°C
Boiling Point 274°C to 276°C
Quantity 5000 g
Beilstein 907218
Merck Index 14,2123
Formula Weight 156.57
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber DG4975990
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.