Learn More
Medchemexpress LLC 3-chlorobenzoic acid-d4 | 2098655-72-0 | MFCD27992341 | >98.0% | 160.59 g/mol | C7HD4ClO2 | 5 MG
SDP
Catalog No. 5000281110
Encompass_Preferred
3-Chlorobenzoic acid-d4 is the deuterated analogue of 3-chlorobenzoic acid used as an internal standard and tracer in analytical chemistry and drug-development studies. Four hydrogen atoms are replaced by deuterium to produce a predictable mass shift for use in mass spectrometry; the substance is supplied as a documented solid with an assigned CAS number for unambiguous identification.
  • Deuterium labeling (d4) for clear mass shift in MS.
  • Suitable as internal standard for quantitative LC-MS/MS assays.
  • Enables metabolic tracing and pharmacokinetic studies.
  • Provided with CAS number and supporting documentation.
  • Available in small quantities ideal for analytical workflows.
Catalog No. 50-002-81110 Supplier Medchemexpress LLC Supplier No. HYW4411745MG
Add to Cart
Add to Cart
3-Chlorobenzoic acid-d4 is the deuterated analogue of 3-chlorobenzoic acid used as an internal standard and tracer in analytical chemistry and drug-development studies. Four hydrogen atoms are replaced by deuterium to produce a predictable mass shift for use in mass spectrometry; the substance is supplied as a documented solid with an assigned CAS number for unambiguous identification.
  • Deuterium labeling (d4) for clear mass shift in MS.
  • Suitable as internal standard for quantitative LC-MS/MS assays.
  • Enables metabolic tracing and pharmacokinetic studies.
  • Provided with CAS number and supporting documentation.
  • Available in small quantities ideal for analytical workflows.