Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorobenzotrifluoride 90.0+%, TCI America™
SDP

Catalog No. C030425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C030425G 25 g
C0304500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C030425G Supplier TCI America Supplier No. C030425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 98-15-7
Molecular Formula C7H4ClF3
Molecular Weight (g/mol) 180.554
MDL Number MFCD00000594
InChI Key YTCGOUNVIAWCMG-UHFFFAOYSA-N
Synonym 3-chlorobenzotrifluoride, 1-chloro-3-trifluoromethyl benzene, m-chlorobenzotrifluoride, benzene, 1-chloro-3-trifluoromethyl, 3-chloro-alpha,alpha,alpha-trifluorotoluene, 3-chlorotrifluoromethylbenzene, m-trifluoromethylphenyl chloride, meta trifluoromethyl chlorobenzene, 3-chlorotrifluorotoluene, ccris 2810
PubChem CID 7374
IUPAC Name 1-chloro-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)Cl)C(F)(F)F

Specifications

Color Yellow
Boiling Point 139°C
Quantity 25 g
UN Number 2234
Formula Weight 180.55
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzotrifluoride
EINECSNumber (3)-0053
RTECSNumber XS9142000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.