Learn More
Medchemexpress LLC 3-Chlorogentisyl alcohol | 32744-80-2 | C7H7ClO3 | 5 MG
SDP
Catalog No. 5000375887
Encompass_Preferred
3-Chlorogentisyl alcohol is a potent E. coli β-glucuronidase inhibitor. It exhibits antiproliferative activity and holds potential for research in anti-cancer and anti-inflammatory therapies.
  • Potent β-glucuronidase inhibitor
  • Exhibits antiproliferative activity
  • Potential for anti-cancer research
  • Potential for anti-inflammatory research
Catalog No. 50-003-75887 Supplier Medchemexpress LLC Supplier No. HY1511685MG
3-Chlorogentisyl alcohol is a potent E. coli β-glucuronidase inhibitor. It exhibits antiproliferative activity and holds potential for research in anti-cancer and anti-inflammatory therapies.
  • Potent β-glucuronidase inhibitor
  • Exhibits antiproliferative activity
  • Potential for anti-cancer research
  • Potential for anti-inflammatory research