Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Chloroindole | 16863-96-0 | >98.4% | C8H6ClN | 250 MG
SDP

Catalog No. 5000358372
Encompass_Preferred

3-Chloroindole is a natural product derived from marine sources, specifically found in Ptychodera flava. It is classified as an indole alkaloid, and it is also noted as being isolated from microorganisms such as Hygrophorus paupertinus. This compound is intended for research use only.

  • Origin: natural product derived from marine sources and microorganisms
  • Appearance: solid
  • Color: yellow to brown
  • Molecular formula: C8H6ClN
  • Molecular weight: 151.60
  • Storage (solid): -20°C, sealed storage, away from moisture
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: 100 mg/mL (659.65 mM) with ultrasonic assistance, recommends using newly opened DMSO due to hygroscopic nature

Catalog No. 50-003-58372 Supplier Medchemexpress LLC Supplier No. 250MGHY65119
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Chloroindole is a natural product derived from marine sources, specifically found in Ptychodera flava. It is classified as an indole alkaloid, and it is also noted as being isolated from microorganisms such as Hygrophorus paupertinus. This compound is intended for research use only.

  • Origin: natural product derived from marine sources and microorganisms
  • Appearance: solid
  • Color: yellow to brown
  • Molecular formula: C8H6ClN
  • Molecular weight: 151.60
  • Storage (solid): -20°C, sealed storage, away from moisture
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: 100 mg/mL (659.65 mM) with ultrasonic assistance, recommends using newly opened DMSO due to hygroscopic nature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.