Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Chloromethyl)-1,2,4-oxadiazole, Technical Grade, Thermo Scientific™

Catalog No. SEW00285EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SEW00285EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW00285EA Supplier Thermo Scientific Chemicals Supplier No. SEW00285EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51791-12-9
Molecular Formula C3H3ClN2O
Molecular Weight (g/mol) 118.52
MDL Number MFCD00052097
InChI Key YSNKGJCEHOJIDK-UHFFFAOYSA-N
PubChem CID 2735765
IUPAC Name 3-(chloromethyl)-1,2,4-oxadiazole
SMILES ClCC1=NOC=N1

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl, chloromethyl 1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl
Formula Weight 118.52
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(Chloromethyl)-1, 2, 4-oxadiazole
Hazard Category Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.