Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Chloroperbenzoic Acid

Catalog No. 273031500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
10 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-303-1500G 500 g
27-303-11KG 1 kg
27-303-125G 25 g
27-303-110KG 10 kg
27-303-1100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 27-303-1500G Supplier Sigma Aldrich Supplier No. 273031500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Chloroperbenzoic Acid

Specifications

CAS 937-14-4
Melting Point 69°C to 71°C
Molecular Formula C7H5ClO3
Linear Formula ClC6H4CO3H
MDL Number MFCD00002127
Quantity 500 g
Synonym meta-Chloroperbenzoic acid; 3-Chloroperoxybenzoic acid; MCPBA
Molecular Weight (g/mol) 172.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.