Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chlorophenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. I011925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I011925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I011925G Supplier TCI America Supplier No. I011925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Melting Point -4°C
CAS 2909-38-8
Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Molecular Formula C7H4ClNO
MDL Number MFCD00002016
UN Number 2206
Synonym 3-chlorophenyl isocyanate, m-chlorophenyl isocyanate, benzene, 1-chloro-3-isocyanato, 3-chlorophenylisocyanate, 3-chloroisocyanatobenzene, isocyanic acid, m-chlorophenyl ester, m-chlorfenylisokyanat, isocyanic acid 3-chlorophenyl ester, m-chlorfenylisokyanat czech, 3-chlorobenzenisocyanate
Boiling Point 204°C
InChI Key HHIRBXHEYVDUAM-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)N=C=O
IUPAC Name 1-chloro-3-isocyanatobenzene
Molecular Weight (g/mol) 153.565
PubChem CID 17968
Formula Weight 153.57
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Chlorophenyl Isocyanate
Show More Show Less
RTECSNumber NQ8560000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.