Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-chlorophenylacetic acid, 99+%

Catalog No. AC109840100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109840100 10 g Glass bottle
AC109840500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109840100 Supplier Thermo Scientific Chemicals Supplier No. 109840100

CAS: 1878-65-5 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-65-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004332
InChI Key WFPMUFXQDKMVCO-UHFFFAOYSA-N
Synonym 3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
PubChem CID 15879
IUPAC Name 2-(3-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Cl)=C1

Specifications

Melting Point 77°C to 80°C
Color White
CAS Min % 99.0
CAS Max % 100.0
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Shiny Flakes and Chunks
Assay Percent Range 99+%
Linear Formula ClC6H4CH2CO2H
Beilstein 09, II, 306
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol
Formula Weight 170.6
Percent Purity 99+%
Chemical Name or Material 3-Chlorophenylacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 217-520-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.