Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Chlorophenylmagnesium Bromide

Catalog No. 56372250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-372-250ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 56-372-250ML Supplier Sigma Aldrich Supplier No. 56372250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Chlorophenylmagnesium bromide (o-chlorophnylmagnesium bromide) is a Grignard reagent that can be used to synthesize: 2,3-Di(o-chlorophenyl)buta-1,3-dienes by reacting with 1,4-dimethoxy 2-butyne in the presence of a copper(I) salt. 2-Bromo-5-chlorothiophen-3-yl-(3-chlorophenyl)methanol by reacting with 2-bromo-5-chlorothiophene-3-carbaldehyde. 2-(3-Chlorophenyl)-1-phenyl-1H-inden-1-ol from 2- benzoylbenzonitrile as a starting a material.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 15112-89-7
Melting Point -90°C
Density 0.838 g/mL at 25°C
Boiling Point 142°C
Molecular Formula C6H19N3Si
Linear Formula ((CH3)2N)3SiH
MDL Number MFCD00048006
Quantity 50 mL
Molecular Weight (g/mol) 161.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.