Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-chloropropane-1,2-diol dipalmitate | 51930-97-3 | MFCD09840071 | 95.0% | 587.36 g·mol-1 | C35H67ClO4 | 5 MG
SDP

Catalog No. 5000284138
Encompass_Preferred

3-Chloropropane-1,2-diol dipalmitate is a long-chain fatty acid diester used as an analytical standard and tracer in lipid research and related biochemical assays. It is provided in small, high-purity quantities suitable for method development, mass spectrometry, and tracer studies in research laboratories.

  • Chemical identity: 3-chloropropane-1,2-diol dipalmitate (CAS 51930-97-3).
  • Molecular weight: 587.36 g·mol-1; formula: C35H67ClO4.
  • Available pack sizes: small milligram quantities (commonly 5 mg to 500 mg).
  • Labeled isotopologue available for tracer studies.
  • Recommended storage: -20°C under nitrogen; in solvent, -80°C (6 months) or -20°C (1 month).
  • Typical applications: analytical standard, tracer, and lipid research reagent.

Catalog No. 50-002-84138 Supplier Medchemexpress LLC Supplier No. HYW5858195MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Chloropropane-1,2-diol dipalmitate is a long-chain fatty acid diester used as an analytical standard and tracer in lipid research and related biochemical assays. It is provided in small, high-purity quantities suitable for method development, mass spectrometry, and tracer studies in research laboratories.

  • Chemical identity: 3-chloropropane-1,2-diol dipalmitate (CAS 51930-97-3).
  • Molecular weight: 587.36 g·mol-1; formula: C35H67ClO4.
  • Available pack sizes: small milligram quantities (commonly 5 mg to 500 mg).
  • Labeled isotopologue available for tracer studies.
  • Recommended storage: -20°C under nitrogen; in solvent, -80°C (6 months) or -20°C (1 month).
  • Typical applications: analytical standard, tracer, and lipid research reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.