Learn More
Medchemexpress LLC 3-chloropropane-1,2-diol dipalmitate | 51930-97-3 | MFCD09840071 | 98.0% | 587.36 g/mol | C35H67ClO4 | 50 MG
SDP
Catalog No. 5000284432
Encompass_Preferred
3-Chloropropane-1,2-diol dipalmitate is a lipid diester supplied as a characterized reference standard for analytical and tracer applications. It is provided as a white to off-white solid with a defined molecular formula and molecular weight, and is intended for quantitative analysis and method development in biochemical and analytical workflows.
  • Used as an analytical standard and tracer for NMR, GC-MS, and LC-MS analyses.
  • White to off-white solid suitable for reference use.
  • Stable when stored under nitrogen; recommended storage in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Well characterized with molecular formula and molecular weight for confident identification.
  • Available in small mg quantities appropriate for analytical assays.
Catalog No. 50-002-84432 Supplier Medchemexpress LLC Supplier No. HYW58581950MG
3-Chloropropane-1,2-diol dipalmitate is a lipid diester supplied as a characterized reference standard for analytical and tracer applications. It is provided as a white to off-white solid with a defined molecular formula and molecular weight, and is intended for quantitative analysis and method development in biochemical and analytical workflows.
  • Used as an analytical standard and tracer for NMR, GC-MS, and LC-MS analyses.
  • White to off-white solid suitable for reference use.
  • Stable when stored under nitrogen; recommended storage in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Well characterized with molecular formula and molecular weight for confident identification.
  • Available in small mg quantities appropriate for analytical assays.