Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-chloropropane-1,2-diol dipalmitate | 51930-97-3 | MFCD09840071 | 98.3% | 587.36 g/mol | C35H67ClO4 | 100 MG
SDP

Catalog No. 5000284273
Encompass_Preferred

3-Chloropropane-1,2-diol dipalmitate is a chlorinated glycerol diester used as an analytical reference standard and tracer in mass spectrometry and NMR workflows. It is described as a free fatty acid derivative composed mainly of diesters and is supplied with documentation for handling and storage to support reproducible quantitative analysis.

  • Provides a well-characterized standard for NMR, GC-MS, and LC-MS applications.
  • High-purity material with batch-specific certificate of analysis available.
  • Molecular formula C35H67ClO4 and molecular weight 587.36 g/mol.
  • Offered in multiple small pack sizes suitable for analytical use.
  • Storage recommendations provided to preserve stability in solid and solution states.

Catalog No. 50-002-84273 Supplier Medchemexpress LLC Supplier No. HYW585819100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Chloropropane-1,2-diol dipalmitate is a chlorinated glycerol diester used as an analytical reference standard and tracer in mass spectrometry and NMR workflows. It is described as a free fatty acid derivative composed mainly of diesters and is supplied with documentation for handling and storage to support reproducible quantitative analysis.

  • Provides a well-characterized standard for NMR, GC-MS, and LC-MS applications.
  • High-purity material with batch-specific certificate of analysis available.
  • Molecular formula C35H67ClO4 and molecular weight 587.36 g/mol.
  • Offered in multiple small pack sizes suitable for analytical use.
  • Storage recommendations provided to preserve stability in solid and solution states.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.